Key Documents
Z332984
BRAND® pipette tips, Tip-Set, non-sterile
About This Item
UNSPSC Code:
41121600
sterility:
non-sterile
feature:
autoclavable
DE-M marking
packaging:
case of 480
rack of 96
material
colorless tip
gray rack
sterility
non-sterile
feature
autoclavable
DE-M marking
packaging
case of 480
rack of 96
General description
Combination of 4 loaded tip-trays (2 Tip-Racks) and 1 filled Tip-Box PC (for 1000 μL and 1250 μL 1 filled Tip-Box N)
  • High-quality PP (free from DiHEMDA and oleamide)
  • Weight reduction of the 200 μL and 1000 μL tips to reduce weight of plastic waste
  • Graduations for a quick volume check
  • BIO-CERT products are free from DNA, RNase and ATP.
  • Autoclavable at 121°C / 250°F (2 bar), acc. DIN EN 285
  • Environmentally friendly packaging
Other Notes
Browse all pipette tips
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)