Key Documents
Z332429
BRAND® discharge tube for Dispensette®
with Luer-lock, for 0.5 mL Dispensette III (not for HF and peroxide)
About This Item
UNSPSC Code:
41121500
manufacturer/tradename
BRAND
L
90 mm
General description
The BRAND® Discharge Tube is designed for use with both the Dispensette® and Seripettor® systems, providing a reliable solution for precise liquid dispensing. This versatile discharge tube includes essential components such as a closure cap, EPDM discharge valve, and sealing ring, ensuring optimal performance and convenience.
Other Notes
BR707928 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)