Z332100
BRAND® Dispensette® III bottle-top dispenser, fixed volume
volume 1 mL, without SafetyPrime
About This Item
Recommended Products
feature
autoclavable
DE-M marking
without SafetyPrime
manufacturer/tradename
BRAND 4700210 EU
BrandTech 4701210 US
volume measuring range
accuracy: 0.5%, precision: 0.1%
volume
1 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Reliable and long lasting through high-grade materials such as ETFE, PTFE, PFA, FEP, Borosilicate glass and Platinum-Iridium
- Safety discharge system. Minimises inadvertent dispensing if discharge tube is improperly installed
- Adjustable dispensing angle. The discharge tube can be positioned in any direction so that the bottle label is always visible. Discharge safety cap
- Telescopic filling tube. Adjusts easily to different size bottles - without measuring and cutting
- Maintenance friendly ball valve
Dispensette III: Ideal for dispening aggressive reagents including strong acids e.g. H3PO4,H2SO4, bases like NaOH, KOH, saline solutions and many solvents (e.g. methanol, acetonitrile).
If fitted with SafetyPrime recirculation valve, avoids reagent waste by recirculating during priming.
Features and Benefits
- Simple: Wide range of applications for organic solvents, acids, alkalis and saline solutions.
- Easy: Minimum operating forces, maximum comfort.
- Efficient: Reliable in challenging continuous operation and with aggressive media.
Linkage
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service