Z329762
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 500 mL, label, Methanol, without venting valve
About This Item
material
green closure
low-density polyethylene bottle
round bottle
translucent bottle
wide-mouth bottle
feature
label Methanol
with cap
without venting valve
packaging
pack of 5 ea
manufacturer/tradename
BRAND 144086
capacity
500 mL
General description
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
Other Notes
Legal Information
accessory
Choose from one of the most recent versions:
Certificates of Analysis (COA)