Z329711
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 250 mL, label, Acetone, without venting valve
About This Item
Recommended Products
material
low-density polyethylene bottle
red closure
round bottle
translucent bottle
wide-mouth bottle
feature
label Acetone
with cap
without venting valve
packaging
pack of 5 ea
manufacturer/tradename
BRAND 144079
capacity
250 mL
General description
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
Other Notes
Legal Information
accessory
Choose from one of the most recent versions:
Certificates of Analysis (COA)