Key Documents
Z329673
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 250 mL, label, Isopropanol, without venting valve
About This Item
UNSPSC Code:
41103209
NACRES:
NB.17
material
blue closure
low-density polyethylene bottle
round bottle
translucent bottle
wide-mouth bottle
feature
label Isopropanol
with cap
without venting valve
packaging
pack of 5 ea
manufacturer/tradename
BRAND 144081
capacity
250 mL
General description
Flexible, high-quality wash bottles with integral wash steam and sealing cone in the closure. Large opening for convenient filling. Label with designation of contents and general safety statements (with CAS no. and NFPA codes).
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
Other Notes
BR144081 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)