Key Documents
Z329320
BRAND® magnetic stirring bar, PTFE, triangular
L 20 mm, bar diam. 8 mm
About This Item
UNSPSC Code:
41103806
eCl@ss:
32100100
material
triangular PTFE-coated
packaging
pack of 10
manufacturer/tradename
BRAND
L
20 mm
bar diam.
8 mm
General description
Wedge shaped. Ideal for dissolving solids and mixing of sediments due to scraping action. Angular surface creates high turbulence even at low speeds.
Other Notes
BR137807 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)