Z328111
BRAND® Transferpette® 8-channel, digital adjustable pipette
volume 5-50 μL, accuracy: 0.8%, precision: 0.4%
About This Item
feature
8-channel
electronic: no
recommended tips 2-200 μL & 5-300 μL
* 5-100 μL & 5-200μL
subdivision 0.1 μL
manufacturer/tradename
BrandTech 703606
volume measuring range
5-50 μL, accuracy: 0.8%, precision: 0.4%
volume
50 μL
General description
Special features: FKM V-rings and a stepped tip ejector significantly reduce the ejection force, easy to maintain with replaceable individual tip cones and seals, universal tip cones - fit all convential pipette tips, corrosion-resistant piston, Easy Calibration Technique: No tools needed to make adjustments, the freely rotatble manifold can be entirely autoclaved at 121°C, separate ejector function.
* Filter tips
Packaging
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)