Z328103
BRAND® Transferpette® 8-channel, digital adjustable pipette
volume 2.5-25 μL, accuracy: 1.0%, precision: 0.6%
About This Item
feature
8-channel
electronic: no
recommended tips 2-200 μL & 5-300 μL
* 5-100 μL & 5-200 μL
subdivision 0.1 μL
manufacturer/tradename
BrandTech 703604
volume measuring range
2.5-25 μL, accuracy: 1.0%, precision: 0.6%
volume
25 μL
General description
Special features: FKM V-rings and a stepped tip ejector significantly reduce the ejection force, easy to maintain with replaceable individual tip cones and seals, universal tip cones - fit all convential pipette tips, corrosion-resistant piston, Easy Calibration Technique: No tools needed to make adjustments, the freely rotatble manifold can be entirely autoclaved at 121°C, separate ejector function.
* Filter tips
Packaging
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)