Z328081
BRAND® Transferpette® 8-channel, digital adjustable pipette
volume 2-20 μL, accuracy: 1.0%, precision: 0.6%
About This Item
Recommended Products
feature
8-channel
electronic: no
recommended tips Nano-cap, 20 μL
* 0.5-10 μL
subdivision 0.1 μL
manufacturer/tradename
BRAND 703602
volume measuring range
2-20 μL, accuracy: 1.0%, precision: 0.6%
volume
20 μL
General description
Special features: FKM V-rings and a stepped tip ejector significantly reduce the ejection force, easy to maintain with replaceable individual tip cones and seals, universal tip cones - fit all convential pipette tips, corrosion-resistant piston, Easy Calibration Technique: No tools needed to make adjustments, the freely rotatble manifold can be entirely autoclaved at 121°C, separate ejector function.
* Filter tips
Packaging
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)