Z325503
BRAND® SILBERBRAND ETERNA graduated pipette, calibrated to deliver (TD, EX)
capacity 2 mL
About This Item
material
AR-Glass
feature
class B
graduation
subdivision 0.1 mL
supplementary to DIN range
manufacturer/tradename
BRAND 27074
capacity
2 mL
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)