Z324795
BRAND® SILBERBRAND ETERNA volumetric pipettes, calibrated to deliver
capacity 1 mL
About This Item
material
AR-Glas®
agency
according to DIN 12 690
feature
class B (callibrated to deliver)
class B
one-mark
packaging
pack of 12 ea
manufacturer/tradename
BRAND 29502
volume measuring range
1 mL, accuracy: 0.010 mL
capacity
1 mL
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.