Z323217
BRAND® wide-mouth bottles with screw cap, PTFE
capacity 50 mL
About This Item
Recommended Products
material
PTFE bottle
round bottle
white bottle
wide-mouth bottle
feature
cap
manufacturer/tradename
BRAND 130528
capacity
50 mL
diam. × H
43 mm × 76 mm
neck I.D.
25 mm
General description
- BRAND® wide-mouth bottles are ideal for safe transport and storing of liquids and powders.
- Polytetrafluoroethylene (PTFE) material offers high chemical and thermal resistance.
- Sturdy, strong-walled designed bottles include a screw cap for sample protection.
- Wide-mouth facilitates easy access to samples.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)