Key Documents
Z317217
Eppendorf® Biopur® Safe-Lock microtubes
volume 1.5 mL
About This Item
UNSPSC Code:
41121702
NACRES:
NB.11
Product Z317217 is not currently sold in your country. Contact Technical Service
material
colorless tube
polypropylene
sterility
sterile
feature
graduated
individually wrapped
packaging
pack of 100 pieces (individually sealed)
manufacturer/tradename
Eppendorf® 0030 121.589
availability
not available in USA and Canada
volume
1.5 mL
foreign activity
DNA, Rnase, ATP and pyrogen, certified free
Application
Eppendorf® Biopur® Safe-Lock microtubes have been used for the isolation of total mRNA from cells.[1] They have been used for the stimulation of the whole blood at 37°C and 5% CO2. Subsequently, they have been used to store the samples at -20°C.[2]
For molecular biology, PCR, pharmaceutical and cell culture applications.
Preparation Note
Manufactured in an automated production under clean-room conditions.
Legal Information
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Impaired genome encapsidation restricts the in vitro propagation of human parvovirus B19.
Wolfisberg R, et al.
J. Virol. Methods, 193, 215-225 (2013)
Holi colours contain PM10 and can induce pro-inflammatory responses.
Bossmann K, et al.
Journal of Occupational Medicine and Toxicology (London, England), 11, 42-42 (2016)