Key Documents
Z316156
Eppendorf® PCR tubes
capacity 0.2 mL
About This Item
UNSPSC Code:
41103037
NACRES:
NB.22
Pricing and availability is not currently available.
material
cap (Lid)
colorless tube
polypropylene
feature
RNase and DNase free
packaging
pkg of 960 (120 × 8 tube strips)
manufacturer/tradename
Eppendorf® 0030 124.359
availability
not available in USA and Canada
technique(s)
PCR: suitable
capacity
0.2 mL
General description
Thin-wall PCR tubes with a contamination shield at the front edge of the lid to prevent accidental contact with the inner surface when the tube is opened.
Matt writing surface on lid
Chem/Phys Resistance
Autoclavable
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)