Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z315672

Eppendorf® Varispenser® Plus

volume 20-100 mL, resolution: 1.0 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

description

precision ± 0.1%
provided with adapters for 32, 38 and 40 mm threads.
telescopic intake tube allows use with short or tall bottles.

feature

fits 45 mm bottle thread

manufacturer/tradename

Eppendorf® 4961 000.063

volume measuring range

20-100 mL, resolution: 1.0 mL

Looking for similar products? Visit Product Comparison Guide

General description

Bottle-top dispensers with PTFE valve blocks and pistons and a Pt-lr spring bolosilicate ball valve for high resistance to aggressive chemicals. Modular construction for simple cleaning and maintenance.

Safety valve for priming without loss of reagent and fine adjustment of volume.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Varispenser is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service