Z315648
Eppendorf® Varispenser® Plus
volume 2-10 mL, accuracy: 0.5%, resolution: 0.20 mL
About This Item
Recommended Products
description
precision ± 0.1%
provided with adapters for 28, 40 and 45 mm threads.
telescopic intake tube allows use with short or tall bottles.
feature
fits 32 mm bottle thread
manufacturer/tradename
Eppendorf® 4961 000.039
volume measuring range
2-10 mL, accuracy: 0.5%, resolution: 0.20 mL
Looking for similar products? Visit Product Comparison Guide
General description
Safety valve for priming without loss of reagent and fine adjustment of volume.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service