Key Documents
Z314870
Hirschmann® akku-drive® control module
Dispenser Module
About This Item
UNSPSC Code:
41121500
description
dispensing speed= 2 mL/sec (using 20 mL base unit)
dispensing speed= 5 mL/sec (using 50 mL base unit)
repetitions= 999 (at intervals of 1-999 sec)
manufacturer/tradename
Hirschmann 9560011
volume measuring range
0.1-200 mL (using 20 mL base unit)
1-500 mL (using 50 mL base unit)
General description
Snaps onto the front of the akku-drive base unit and controls dispensing functions to the specifications given. Accuracy and reproducibility based on a nominal volume 20 ml deionised water.
Legal Information
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
akku-drive is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)