Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y4250

Sigma-Aldrich

Yeast Extract

BioReagent, suitable for plant cell culture

Synonym(s):

Select Yeast Extract

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
42040000

product line

BioReagent

technique(s)

cell culture | plant: suitable

solubility

H2O: 10% (remains clear after heating to 40°C)

application(s)

agriculture

InChI

1S/C19H14O2/c20-18(21)11-17-15-8-4-3-7-14(15)16-10-9-12-5-1-2-6-13(12)19(16)17/h1-10,17H,11H2,(H,20,21)

InChI key

GQNBDGXKDJSVGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For general bacteriological use with a variety of microorganisms.
Yeast extract is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.

Other Notes

Spray dried, autolyzed yeast
Water soluble portion of autolyzed yeast with intact B-complex vitamins. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F T Sabie et al.
Mycopathologia, 119(3), 147-156 (1992-09-01)
A yeast-mycelium (Y-M) transition in Candida albicans was induced by exogenous yeast extract, adenosine, adenosine 5'-monophosphate (AMP), adenosine 5'-diphosphate (ADP), adenosine 3':5' cyclic monophosphate (cAMP) and its analogue N6, O2'-dibutyryl adenosine 3':5'-cyclic monophosphate (dbcAMP) in defined liquid medium at 25
Effect of glucose addition and N sources in defined media on fibrolytic activity profiles of Neocallimastix sp. YQ1 grown on corn stover.
Yang HJ, Yue Q.
Journal of animal physiology and animal nutrition (2011)
Jae Su Kim et al.
Journal of microbiology and biotechnology, 21(6), 604-612 (2011-07-01)
Beauveria bassiana SFB-205 supernatant can effectively control cotton aphid populations, which is closely associated with its chitinase activity. The present work extends to optimizing a culture medium to produce more efficacious supernatant in flask conditions, followed by scale-up in 7
W J VanDusen et al.
Biotechnology progress, 13(1), 1-7 (1997-01-01)
We have described a method to reliably measure the free adenine content of yeast extract powders or the adenine concentrations found in chemically-defined and complex fermentation samples. This method relies on the selective precolumn derivatization of adenine with chloroacetaldehyde to
Jiang-Lin Zhao et al.
Pharmacognosy magazine, 10(39), 234-240 (2014-09-12)
Tartary buckwheat (Fagopyrum tataricum), an excellent edible and medicinal crop, has been widely used as a daily diet and traditional medicine for a long time. The major functional components of Fagopyrum tataricum have been demonstrated to be flavonoids (i.e. rutin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service