Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

XNABRE

Sigma-Aldrich

REDExtract-N-Amp Blood PCR Kit

sufficient for 1000 extractions, sufficient for 5000 amplifications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for 1000 extractions
sufficient for 5000 amplifications

feature

hotstart

color

red

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The REDExtract-N-Amp Blood PCR Kits contain all of the reagents necessary to rapidly extract host genomic DNA from whole blood and amplify targets of interest by PCR. This novel extraction system eliminates the need for any type of purification, organic extraction, centrifugation, heating, filtration, or alcohol precipitation. The kit also includes a PCR ReadyMix, that uses an antibody based hot start and is specially formulated for amplification directly from the extract. The REDExtract-N-Amp Blood PCR Kit contains a tracking dye that allows for convenient direct loading of PCR reactions onto agarose gels for analysis.

Genomic DNA is extracted from 10 μL of whole blood by simply adding the Extraction Solution and incubating for 5 minutes at room temperature. The Neutralization Solution is added to the extract to counteract inhibitory substances prior to PCR. A portion of the DNA extract is then added to the specially formulated PCR Mix.
The REDExtract-N-Amp Blood PCR Kits contain all of the reagents necessary to rapidly extract host genomic DNA from whole blood and amplify targets of interest by direct PCR (polymerase chain reaction). This novel extraction system eliminates the need for any type of purification, organic extraction, centrifugation, heating, filtration or alcohol precipitation. The kit also includes a PCR ReadyMix, specially formulated for amplification directly from the extract. This formulation uses an antibody based hot start, for specific amplification. The REDExtract-N-Amp Blood PCR Kit contains a tracking dye that allows for convenient direct loading of PCR reactions onto agarose gels for analysis.

Application

REDExtract-N-Amp Blood PCR Kit has been used to extract genomic DNA from blood samples, peripheral blood leukocytes, parasites and endocervical swab eluates of mice.[1][2][3][4][5][6] It has also been used to perform polymerase chain reaction (PCR) using oligonucleotide primers.[7]

Features and Benefits

  • Efficient 8 minute prep allows greater speed and throughput
  • No need for organic extraction, centrifugation or alcohol precipitation
  • Hot start antibody for highly specific PCR amplification of genomic DNA
  • Can be used with whole blood or blood cards
  • Extract stable at 4 °C for at least 6 months
  • Compatible with any format (single tube, 96-well, etc.)
  • Simple, 3 step procedure with no special equipment required
  • REDExtract-N-Amp requires no loading buffers or tracking dyes required for gel analysis
  • PCR products are suitable for direct cloning and sequencing

Principle

Genomic DNA is extracted from 10 μl of whole blood by simply adding the Extraction Solution and incubating for 5 minutes at room temperature. The Neutralization Solution is added to the extract to counteract inhibitory substances prior to PCR. A portion of the DNA extract is then added to the specially formulated PCR ReadyMix.

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.
Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
REDExtract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofia Lejon Crottet et al.
Transfusion and apheresis science : official journal of the World Apheresis Association : official journal of the European Society for Haemapheresis, 50(2), 169-174 (2014-04-01)
Starting in 2013, blood donors must be tested at least using: (1) one monoclonal anti-D and one anti-CDE (alternatively full RhCcEe phenotyping), and (2) all RhD negative donors must be tested for RHD exons 5 and 10 plus one further
Olivier A E Sparagano et al.
Annals of the New York Academy of Sciences, 1081, 505-508 (2006-12-01)
Our study is based on the collection of blood and ticks from sheep in Iran and Italy. Polymerase chain reaction (PCR) testing was performed to target the 18S rRNA gene and RLB was performed using previously published probes. In Italy
Dawn M Reeves et al.
Antimicrobial agents and chemotherapy, 51(7), 2646-2648 (2007-05-16)
Mice treated with antibiotics early or late after active infection had resolved were examined for chlamydial DNA in endocervical swabs. The early eradication of infection limited oviduct pathology, despite the continued detection of chlamydial DNA by nested PCR. Late antibiotic
Anna K Nowińska et al.
Molecular vision, 17, 2333-2342 (2011-09-17)
To analyze genotype-phenotype correlation in patients originating from Polish population with the transforming growth factor beta induced (TGFBI) corneal dystrophies. Sixty affected and 31 unaffected individuals from 15 unrelated Polish families were included in the study. The clinical diagnosis was
Jun Liu et al.
Eukaryotic cell, 12(12), 1588-1599 (2013-07-23)
Microtubules are versatile biopolymers that support numerous vital cellular functions in eukaryotes. The specific properties of microtubules are dependent on distinct microtubule-associated proteins, as the tubulin subunits and microtubule structure are exceptionally conserved. Highly specialized microtubule-containing assemblies are often found

Articles

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service