Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

X3129

Sigma-Aldrich

XRP44X

≥98% (HPLC)

Synonym(s):

1-(3-Chlorophenyl)-4-((3-methyl-1-phenyl-1H-pyrazol-5-yl)carbonyl)piperazine, [4-(3-Chlorophenyl)-1-piperazinyl](3-methyl-1-phenyl-1H-pyrazol-5-yl)-methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClN4O
CAS Number:
Molecular Weight:
380.87
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

originator

Sanofi Aventis

storage temp.

room temp

SMILES string

Cc1cc(C(=O)N2CCN(CC2)c3cccc(Cl)c3)n(n1)-c4ccccc4

InChI

1S/C21H21ClN4O/c1-16-14-20(26(23-16)18-7-3-2-4-8-18)21(27)25-12-10-24(11-13-25)19-9-5-6-17(22)15-19/h2-9,14-15H,10-13H2,1H3

InChI key

NPHPNBGYPKBDDB-UHFFFAOYSA-N

Biochem/physiol Actions

XRP44X is a Ras-Net (Elk-3) pathway inhibitor; tubulin polymerization inhibitor Colchicine binding site.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juha Mehtonen et al.
Genome medicine, 12(1), 99-99 (2020-11-22)
Tight regulatory loops orchestrate commitment to B cell fate within bone marrow. Genetic lesions in this gene regulatory network underlie the emergence of the most common childhood cancer, acute lymphoblastic leukemia (ALL). The initial genetic hits, including the common translocation
Lisa P Elia et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(17), 3332-3344 (2019-01-31)
Deficient progranulin levels cause dose-dependent neurological syndromes: haploinsufficiency leads to frontotemporal lobar degeneration (FTLD) and nullizygosity produces adult-onset neuronal ceroid lipofuscinosis. Mechanisms controlling progranulin levels are largely unknown. To better understand progranulin regulation, we performed a genome-wide RNAi screen using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service