Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

X1504

Sigma-Aldrich

Anti-XPA (N-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-XP1, Anti-XPAC, Anti-Xeroderma pigmentosum, complementary group A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 35 kDa (doublet)

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using HEK-293T cell lysates
indirect immunofluorescence: 5-10 μg/mL using A549 cells fixed with paraformaldehyde/Triton
western blot: 0.5-1 μg/mL using Jurkat cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... XPA(7507)

Immunogen

synthetic peptide corresponding to amino acids 15-33 of human XPA, conjugated to KLH via a C-terminal added cysteine residue. The immunizing peptide differs from the rat sequence in three amino acids.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Target description

XPA (N-terminal), xeroderma pigmentosum, complementation group A, encodes a zinc finger protein involved in DNA excision repair. The encoded protein is part of the NER (nucleotide excision repair) complext which is responsible for repair of UV radiation-i

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iltaeg Cho et al.
PLoS genetics, 9(4), e1003407-e1003407 (2013-05-03)
The Cockayne syndrome complementation group B (CSB) protein is essential for transcription-coupled DNA repair, and mutations in CSB are associated with Cockayne syndrome--a devastating disease with complex clinical features, including the appearance of premature aging, sun sensitivity, and numerous neurological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service