WPB01549WA
Whirl-Pak® High Temperature Bag
1627 mL capacity(55 oz)
Synonym(s):
autoclave
All Photos(1)
About This Item
UNSPSC Code:
24111500
sterility
sterile
packaging
pkg of 500 ea
manufacturer/tradename
B01549WA (Nasco)
W × L × thickness
19 cm × 30 cm × 0.076 mm
capacity
1627 mL × 55 oz
1627 mL capacity(55 oz)
General description
For hot fill samples with a temperature up to 215° F (102° C), the Whirl-Pak high temperature bag is specially made for this application. The FDA approved film is a co-extrusion of low density polyethylene and nylon that will hold liquids, solids, sauces, gravies, and other samples having a high temperature. The bag is self standing and does not need a rack or special holder to keep it upright, making sample collection safe and easy. The write-on strip provides an easy place to list sample information on the bag. (Do not put bag in boiling water.)
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate. Bags can be frozen to any temperature, even liquid nitrogen - careful handling required after freezing. All bags are sterilized after manufacturing.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate. Bags can be frozen to any temperature, even liquid nitrogen - careful handling required after freezing. All bags are sterilized after manufacturing.
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)