Key Documents
WPB01548WA
Whirl-Pak® 1-Ply Composite Tissue Bag
Synonym(s):
wipers
About This Item
UNSPSC Code:
24111500
packaging
box of 100 ea
manufacturer/tradename
B01548WA (Nasco)
W × L × thickness
11.5 cm × 23 cm × 0.064 mm
capacity
532 mL × 18 oz
532 mL capacity(18 oz)
General description
Includes a 1-ply composite tissue in bottom compartment. To use, tear off the bottom compartment, hydrate tissue with neutralizing buffer, remove using a glove, and collect surface sample. After collection, place tissue inside top of the bag, seal, and return to lab. This is an effective, low-cost alternative to other surface sampling methods.
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)