WPB01478WA
Whirl-Pak® Bag with ScoopBag
532 mL(18 oz), with sterile teaspoon
All Photos(1)
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
sterility
sterile
packaging
pkg of 100 ea
manufacturer/tradename
B01478WA (Nasco)
Whirl-pak®
W × L × thickness
11.5 cm × 23 cm × 0.064 mm
capacity
532 mL(18 oz)
General description
For fast, convenient sampling of a variety of materials. The sterile scoop is already contained inside the bag, eliminating the need to carry two items to collect the sample and minimizing contamination. It is ideal for a wide range of dry products, powders, and other similar materials.
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Customers Also Viewed
Sophie Budge et al.
PloS one, 15(5), e0232541-e0232541 (2020-05-10)
Early infection from enteropathogens is recognised as both a cause and effect of infant malnutrition. Specifically, evidence demonstrates associations between growth shortfalls and Campylobacter infection, endemic across low-income settings, with poultry a major source. Whilst improvements in water, sanitation and