Key Documents
WPB01255WA
Whirl-Pak® Pocket Bag
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
sterility
sterile
packaging
pkg of 500 ea
manufacturer/tradename
B01255WA (Nasco)
parameter
-346-180 °F temp. range (-210 to 82 °C)
W × L × thickness
15 cm × 23 cm × 0.064 mm
capacity
710 mL × 24 oz
710 mL capacity(24 oz)
General description
You′ll find many uses for this bag featuring an outside pocket for paperwork. The pocket keeps important paperwork dry, clean, and separate from the sample.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F (82° C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to -346° F (-210° C). Careful handling required after freezing. All bags are sterilized after manufacturing.
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)