MilliporeSigma
All Photos(5)
Key Documents
WPB01253WA
Whirl-Pak® Trans-Pak® Specimen Bag
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
description
Pocket for paperwork
packaging
pkg of 500 ea
manufacturer/tradename
B01253WA (Nasco)
Trans-Pak®
Whirl-pak®
parameter
<180 °F temp. limit (82 °C)
W × L × thickness
15 cm × 23 cm × 0.064 mm
General description
Are the people transporting specimens in your facility fully protected? A unique outside pouch to separate important paperwork from the inside of the bag and the specimen - eliminating misplaced documents! The specimen transport bag also features:
  • Biohazard symbol required by OSHA (Occupational Safety and Health Administration)
  • Unbreakable polyethylene
  • Easy to handle
  • Positive closure with “PUNCTURE PROOF TABS”
  • Minimum storage and carrying space
Resins used in the extrusion of film are FDA approved. Bags are manufactured in accordance with the requirements for manufacturers of single service containers for milk and milk products. Capacity is measured in fluid ounces when bag is closed and tab folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F (82° C).
Legal Information
Trans-Pak is a registered trademark of The Aristotle Corporation
Whirl-pak is a registered trademark of The Aristotle Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)