Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0414899M5

Sigma-Aldrich

Monoclonal Anti-BRCC2 antibody produced in mouse

clone 3F4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BRCC2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$406.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BLID(414899)

Related Categories

General description

This gene encodes a BH3-like motif containing protein involved in cell death. The encoded protein may induce apoptosis in a caspase-dependent manner. The protein is localized in both the cytoplasm and the mitochondrion. [SH

Immunogen

BRCC2 (NP_001001786, 2 a.a. ~ 108 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VTLLPIEGQEVHFFEILESECVLYTGWIERASGSSIYPEAKARLPLEALLGSNKEPMLPKETVLSLKRYNLGSSAMKRNVPGHVLQRPSYLTRIQVTLLCNSSAEAL

Biochem/physiol Actions

BRCC2 or BH3-like motif containing, cell death inducer acts as a tumor suppressor by modulating the AKT8 virus oncogene cellular homolog (Akt) pathway in breast cancer tissue. It associates with Bcl-2 long isoform (BCL-XL). BRCC2 also stimulates the caspase-dependent mitochondrial cell death pathway.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frequent loss of the BLID gene in early-onset breast cancer.
Cavalli LR
Cytogenetic and genome research, 135(1), 19-24 (2011)
BRCC2 inhibits breast cancer cell growth and metastasis in vitro and in vivo via downregulating AKT pathway.
Li X
Cell Death & Disease, 4, e757-e757 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service