Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0404037M2

Sigma-Aldrich

Monoclonal Anti-HAPLN4 antibody produced in mouse

clone 1C1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BRAL2, Anti-KIAA1926, Anti-hyaluronan and proteoglycan link protein 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HAPLN4(404037)

Related Categories

General description

Hyaluronan and proteoglycan link protein 4 (HAPLN4) is expressed in the brain.

Immunogen

HAPLN4 (NP_075378, 30 a.a. ~ 134 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
QRGRKKVVHVLEGESGSVVVQTAPGQVVSHRGGTIVLPCRYHYEAAAHGHDGVRLKWTKVVDPLAFTDVFVALGPQHRAFGSYRGRAELQGDGPGDASLVLRNVT

Biochem/physiol Actions

Hyaluronan and proteoglycan link protein 4 (HAPLN4) takes part in the development of the extracellular matrix. It also has a role in adhesion and migration of glioma cells. HAPLN4 aids in the association of hyaluronan with other proteins like aggrecan.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hosung Sim et al.
The Journal of biological chemistry, 284(39), 26547-26556 (2009-07-28)
Malignant gliomas have a distinctive ability to infiltrate the brain parenchyma and disrupt the neural extracellular matrix that inhibits motility of axons and normal neural cells. Chondroitin sulfate proteoglycans (CSPGs) are among the major inhibitory components in the neural matrix
Yoko Bekku et al.
Molecular and cellular neurosciences, 24(1), 148-159 (2003-10-11)
The hyaluronan binding chondroitin sulphate proteoglycans, called lecticans, are the abundant extracellular matrix molecules in the developing and/or adult brain. The link proteins (LPs) are also known to be coordinately present in brain. We report here the molecular cloning and
Anna K Kähler et al.
Biological psychiatry, 69(1), 90-96 (2010-10-19)
The Human Natural Killer-1 carbohydrate (HNK-1) is involved in neurodevelopment and synaptic plasticity. Extracellular matrix structures called perineuronal nets, condensed around subsets of neurons and proximal dendrites during brain maturation, regulate synaptic transmission and plasticity. Ten genes of importance for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service