Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

WH0376267M1

Sigma-Aldrich

Monoclonal Anti-RAB15 antibody produced in mouse

clone 1G12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-RAB15, member RAS onocogene family

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB15(376267)

General description

Member of RAS oncogene family, RAB15 is a GTPase expressed in the brain.

Immunogen

RAB15 (AAH40679, 99 a.a. ~ 208 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
IMKWVSDVDEVGDATSLPGCGEGASPGKARRGPDGKANASRKLCLPQPWMKTSGTHQKASRRSLLGIRLMRSRNGRWEESKGSSWRRSMAWTSMKQVPAPTSTLKSHSRV

Biochem/physiol Actions

Member of RAS oncogene family, RAB15, has a role in endocytosis. It is a target of the protein atonal homolog 1 (Atoh1), a neural transcription factor. RAB15 has been linked to neuroblastoma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thi Van Huyen Pham et al.
Oncology reports, 27(6), 2045-2049 (2012-03-20)
Neuroblastoma is an aggressive pediatric tumor that accounts for 15% of cancer-related deaths in children. More than half of high-risk neuroblastoma patients develop tumor relapse that is lethal in most cases. A small population of tumor-initiating cells (TICs), recently identified
David J Strick et al.
Molecular biology of the cell, 16(12), 5699-5709 (2005-10-01)
Sorting endosomes and the endocytic recycling compartment are critical intracellular stores for the rapid recycling of internalized membrane receptors to the cell surface in multiple cell types. However, the molecular mechanisms distinguishing fast receptor recycling from sorting endosomes and slow

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service