Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

WH0221823M1

Sigma-Aldrich

Monoclonal Anti-PRPS1L1 antibody produced in mouse

clone 5E10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-PRPS1, Anti-PRPS3, Anti-PRPSL, Anti-PRSIII, Anti-phosphoribosyl pyrophosphate synthetase 1-like 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5E10, monoclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG3κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Related Categories

General description

This intronless gene is specifically expressed in the testis, and encodes a protein that is highly homologous to the two subunits of phosphoribosylpyrophosphate synthetase encoded by human X-linked genes, PRPS1 and PRPS2. These enzymes convert pyrimidine, purine or pyridine bases to the corresponding ribonucleoside monophosphates. In vitro transcription/translation and site-directed mutagenesis studies indicate that translation of this mRNA initiates exclusively at a non-AUG (ACG) codon. (provided by RefSeq)

Immunogen

PRPS1L1 (NP_787082, 146 a.a. ~ 243 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
YAEPTVLKWIRENIPEWKNCIIVSPDAGGAKRVTSIADQLNVDFALIHKERKKANEVDCIVLVGDVNDRVAILVDDMADTCVTICLAADKLLSAGATR

Biochem/physiol Actions

Phosphoribosyl pyrophosphate synthetase 1-like 1 (PRPS1L1) might be linked to rheumatoid arthritis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew P Morris et al.
BMC proceedings, 3 Suppl 7, S131-S131 (2009-12-19)
Established loci for rheumatoid arthritis (RA), including HLA-DRB1 and PTPN22, do not fully account for the genetic component of susceptibility to the disease. One possible source of as yet undiscovered susceptibility genes are those mediated through effects of rare variants.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service