Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0200095M1

Sigma-Aldrich

Monoclonal Anti-C1orf67 antibody produced in mouse

clone 3C12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-hypothetical protein MGC27277

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

C1orf67 or dynein axonemal heavy chain 14 (DNAH14) is a motor protein. The gene encoding it is localized on human chromosome 1.

Immunogen

MGC27277 (AAH42869, 1 a.a. ~ 189 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDKEETKTKPRLLRYEEKKYEDVKPLETQPAEIAEKETLEYKTVRTFSESLKSEKTEDYLRESIIQQHMVSPEPASLKEKGKSRRKKDQTHACPNVRKARPVSYDRTEPKDDDVIRNIIRLREKLGWQTILPQHSLKYGSSKIAIQKITLKKPLEDDGEFVYCLPRKSPKSLYNPYDLQVINIVGSVKE

Biochem/physiol Actions

C1orf67 associates with cytoskeletal microtubules. It has been linked to embryonic lethality.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosemary Braun et al.
PloS one, 8(3), e55037-e55037 (2013-04-05)
Ovarian cancer remains a significant public health burden, with the highest mortality rate of all the gynecological cancers. This is attributable to the late stage at which the majority of ovarian cancers are diagnosed, coupled with the low and variable
Hanan E Shamseldin et al.
Genome biology, 16, 116-116 (2015-06-04)
Identifying genetic variants that lead to discernible phenotypes is the core of Mendelian genetics. An approach that considers embryonic lethality as a bona fide Mendelian phenotype has the potential to reveal novel genetic causes, which will further our understanding of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service