Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0166614M1

Sigma-Aldrich

Monoclonal Anti-DCAMKL2 antibody produced in mouse

clone 2A5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DCDC3, Anti-DCK2, Anti-DKFZp761I032, Anti-MGC45428, Anti-doublecortin and CaM kinase-like 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DCLK2(166614)

General description

Doublecortin and CaM kinase-like 2 (DCAMKL2) is expressed in the cell body and terminal regions of dendrites and axons. The gene encoding this protein is located on human chromosome 4q31.3.

Immunogen

DCAMKL2 (AAH32726, 348 a.a. ~ 438 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
FRGLKISAHGRSSSNVNGGPELDRCISPEGVNGNRCSESSTLLEKYKIGKVIGDGNFAVVKECIDRSTGKEFALKIIDKAKCCGKEHLIE*

Biochem/physiol Actions

Doublecortin and CaM kinase-like 2 (DCAMKL2) has a role in stabilizing microtubules.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kornsorn Srikulnath et al.
Chromosome research : an international journal on the molecular, supramolecular and evolutionary aspects of chromosome biology, 17(8), 975-986 (2009-11-26)
The butterfly lizard (Leiolepis reevesii rubritaeniata) has the diploid chromosome number of 2n = 36, comprising two distinctive components, macrochromosomes and microchromosomes. To clarify the conserved linkage homology between lizard and snake chromosomes and to delineate the process of karyotypic
Arthur M Edelman et al.
The Journal of biological chemistry, 280(9), 8531-8543 (2004-12-22)
The microtubule (MT)-associated DCX protein plays an essential role in the development of the mammalian cerebral cortex. We report on the identification of a protein kinase, doublecortin kinase-2 (DCK2), with a domain (DC) highly homologous to DCX. DCK2 has MT

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service