Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

WH0161582M9

Sigma-Aldrich

Monoclonal Anti-DYX1C1 antibody produced in mouse

clone 6G1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DYX1, Anti-DYXC1, Anti-EKN1, Anti-FLJ37882, Anti-MGC70618, Anti-dyslexia susceptibility 1 candidate 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DYX1C1(161582)

General description

Dyslexia susceptibility 1 candidate 1 (DYX1C1) is a 420 amino acid protein. The gene encoding it localized on human chromosome 15q21 and consists of 10 exons.[1]

Immunogen

DYX1C1 (NP_570722, 336 a.a. ~ 420 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KLKNLHKAIEDSSKALELLMPPVTDNANARMKAHVRRGTAFCQLELYVEGLQDYEAALKIDPSNKIVQIDAEKIRNVIQGTELKS

Biochem/physiol Actions

Dyslexia susceptibility 1 candidate 1 (DYX1C1) associates with estrogen receptors. It has a role in neuronal migration and has been linked to developmental dyslexia.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Tran et al.
American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics, 162B(2), 146-156 (2013-01-24)
Reading disabilities (RD) have a significant genetic basis and have shown linkage to multiple regions including chromosome 15q. Dyslexia susceptibility 1 candidate gene 1 (DYX1C1) on chromosome 15q21 was originally proposed as a candidate gene with two potentially functional polymorphisms
Shyamala K Venkatesh et al.
Psychiatric genetics, 24(1), 10-20 (2013-12-24)
DYX1C1 has been identified as a susceptible candidate gene for developmental dyslexia (DD); studies in various populations have yielded inconclusive results and the causal allele is unknown in the Indian population. On the basis of the initial association studies and
Gustaf Rosin et al.
BMC cancer, 12, 79-79 (2012-03-02)
The dyslexia candidate gene, DYX1C1, shown to regulate and interact with estrogen receptors and involved in the regulation of neuronal migration, has recently been proposed as a putative cancer biomarker. This study was undertaken to assess the prognostic value and
T C Bates et al.
Molecular psychiatry, 15(12), 1190-1196 (2009-11-11)
The status of DYX1C1 (C15q21.3) as a susceptibility gene for dyslexia is unclear. We report the association of this gene with reading and spelling ability in a sample of adolescent twins and their siblings. Family-based association analyses were carried out

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service