Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0083999M1

Sigma-Aldrich

Monoclonal Anti-KREMEN1 antibody produced in mouse

clone 1E7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FLJ31863, Anti-KREMEN, Anti-KRM1, Anti-kringle containing transmembrane protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KREMEN1(83999)

General description

This gene encodes a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. The encoded protein is a component of a membrane complex that modulates canonical WNT signaling through lipoprotein receptor-related protein 6 (LRP6). It contains extracellular kringle, WSC, and CUB domains. Alternatively spliced transcript variants encoding distinct isoforms have been observed for this gene. (provided by RefSeq)

Immunogen

KREMEN1 (NP_114434, 310 a.a. ~ 391 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
INQAQGFAVLYQAVKEELPQERPAVNQTVAEVITEQANLSVSAARSSKVLYVITTSPSHPPQTVPGSNSWAPPMGAGSHRVE

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service