Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

WH0079813M5

Sigma-Aldrich

Monoclonal Anti-EHMT1 antibody produced in mouse

clone 3G1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-EUHMTASE1, Anti-EuHMTase1, Anti-FLJ12879, Anti-FP13812, Anti-GLP, Anti-KIAA1876, Anti-bA188C12.1, Anti-euchromatic histone-lysine N-methyltransferase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... EHMT1(79813)

General description

Euchromatic histone lysine methyltransferase 1 (EHMT1) is also known as G9a-like protein (GLP). It possesses N-terminal domains that have six ankyrin repeats. The gene encoding EHMT1 is localized on human chromosome 9q34.3.

Immunogen

EHMT1 (NP_079033, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAADEGSAEKQAGEAHMAADGETNGSCENSDASSHANAAKHTQDSARVNPQDGTNTLTRIAENGVSERDSEAAKQNHVTADDFVQTSVIGSNGYILNKPA

Biochem/physiol Actions

Euchromatic histone lysine methyltransferase 1 (EHMT1) mono- and di-methylates histone H3 at the lysine 9 residue. Defects in the gene have been associated with Kleefstra syndrome or 9q34 deletion syndrome. EHMT1 associates with the transcription factor E2F6 and is involved in transcriptional inactivation through chromatin remodeling.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intragenic duplication of EHMT1 gene results in Kleefstra syndrome.
Schwaibold EM
Molecular Cytogenetics (2014)
A Novel Kleefstra Syndrome-associated Variant That Affects the Conserved TPLX Motif within the Ankyrin Repeat of EHMT1 Leads to Abnormal Protein Folding.
Blackburn PR
The Journal of Biological Chemistry (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service