Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0079768M1

Sigma-Aldrich

Monoclonal Anti-C15orf29 antibody produced in mouse

clone 1E7, ascites fluid

Synonym(s):

Anti-FLJ22557, Anti-chromosome 15 open reading frame 29

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
200 μL
$406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1E7, monoclonal

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgMκ

GenBank accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

C15orf29 (AAH26234.1, 1 a.a. ~ 304 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MASETHNVKKRNFCNKIEDHFIDLPRKKISNFTNKNMKEVKKSPKQLAAYINRTVGQTVKSPDKLRKVIYRRKKVHHPFPNPCYRKKQSPGSGGCDMANKENELACAGHLPEKLHHDSRTYLVNSSDSGSSQTESPSSKYSGFFSEVSQDHETMAQVLFSRNMRLNVALTFWRKRSISELVAYLLRIEDLGVVVDCLPVLTNCLQEEKQYISLGCCVDLLPLVKSLLKSKFEEYVIVGLNWLQAVIKRWWSELSSKTEIINDGNIQILKQQLSGLWEQENHLTLVPGYTGNIAKDVDAYLLQLH

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service