Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

WH0079084M1

Sigma-Aldrich

Monoclonal Anti-WDR77 antibody produced in mouse

clone 3F10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-HKMT1069, Anti-MEP50, Anti-MGC2722, Anti-WD repeat domain 77

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F10, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... WDR77(79084)

General description

WDR77 is a component of the 20S PRMT5 (MIM 604045)-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins (see MIM 601061). This modification targets Sm proteins to the survival of motor neurons (SMN) complex (see MIM 600354) for assembly into small nuclear ribonucleoprotein core particles (Friesen et al., 2002 [PubMed 11756452]).[supplied by OMIM

Immunogen

WDR77 (AAH01679, 1 a.a. ~ 342 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MRKETPPPLVPPAAREWNLPPNAPACMERQLEAARYRSDGALLLGASSLSGRCWAGSLWLFKDPCAAPNEGFCSAGVQTEAGVADLTWVGERGILVASDSGAVELWELDENETLIVSKFCKYEHDDIVSTVSVLSSGTQAVSGSKDICIKVWDLAQQVVLSSYRAHAAQVTCVAASPHKDSVFLSCSEDNRILLWDTRCPKPASQIGCSAPGYLPTSLAWHPQQSEVFVFGDENGTVSLVDTKSTSCVLSSAVHSQCVTGLVFSPHSVPFLASLSEDCSLAVLDSSLSELFRSQAHRDFVRDATWSPLNHSLLTTVGWDHQVVHHVVPTEPLPAPGPASVTE

Biochem/physiol Actions

WD repeat domain 77 (WDR77) associates with protein arginine methyltransferase 5 (PRMT5) in a methylosome complex and stimulates its histone methyltransferase activity. It also binds to androgen receptors and modulates its target genes in prostate epithelial cells and prostate cancer.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subcellular localization of p44/WDR77 determines proliferation and differentiation of prostate epithelial cells.
Gao S and Wang Z
PLoS ONE, 7(11), e49173-e49173 (2012)
The PRMT5 arginine methyltransferase: many roles in development, cancer and beyond.
Stopa N
Cellular and Molecular Life Sciences, 72(11), 2041-2059 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service