Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0064327M5

Sigma-Aldrich

Monoclonal Anti-LMBR1 antibody produced in mouse

clone 4A1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ACHP, Anti-C7orf2, Anti-FLJ11665, Anti-limb region 1 homolog (mouse)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMBR1(64327)

General description

This gene encodes a member of the LMBR1-like membrane protein family. Another member of this protein family has been shown to be a lipocalin transmembrane receptor. A highly conserved, cis-acting regulatory module for the sonic hedgehog gene is located within an intron of this gene. Consequently, disruption of this genic region can alter sonic hedgehog expression and affect limb patterning, but it is not known if this gene functions directly in limb development. Mutations and chromosomal deletions and rearrangements in this genic region are associated with acheiropody and preaxial polydactyly, which likely result from altered sonic hedgehog expression. (provided by RefSeq)

Immunogen

LMBR1 (NP_071903, 214 a.a. ~ 295 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SRMFTVMGQLLVKPTILEDLDEQIYIITLEEEALQRRLNGLSSSVEYNIMELEQELENVKTLKTKLERRKKASAWERNLVYP

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service