Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0056832M1

Sigma-Aldrich

Monoclonal Anti-IFNK antibody produced in mouse

clone 1B7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-RP1127J8.1, Anti-interferon, kappa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... IFNK(56832)

General description

This gene encodes a member of the type I interferon family. Type I interferons are a group of related glycoproteins that play an important role in host defenses against viral infections. This protein is expressed in keratinocytes and the gene is found on chromosome 9, adjacent to the type I interferon cluster. (provided by RefSeq)

Immunogen

IFNK (NP_064509, 35 a.a. ~ 129 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VHLRRVTWQNLRHLSSMSNSFPVECLRENIAFELPQEFLQYTQPMKRDIKKAFYEMSLQAFNIFSQHTFKYWKERHLKQIQIGLDQQAEYLNQCL

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service