Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

WH0051156M2

Sigma-Aldrich

Monoclonal Anti-SERPINA10 antibody produced in mouse

clone 1E11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-PZI, Anti-ZPI, Anti-serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

General description

Serpin family A member 10 (SERPIN10) or protein Z-dependent inhibitor is a single-chain glycoprotein which is expressed in the liver. The gene encoding it is localized on human chromosome 14q32.13.

Immunogen

SERPINA10 (AAH22261, 22 a.a. ~ 444 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LAPSPQSPETPAPQNQTSRVVQAPKEEEEDEQEASEEKASEEEKAWLMASRQQLAKETSNFGFSLLRKISMRHDGNMVFSPFGMSLAMTGLMLGATGPTETQIKRGLHLQALKPTKPGLLPSLFKGLRETLSRNLELGLTQGSFAFIHKDFDVKETFFNLSKRYFDTECVPMNFRNASQAKRLMNHYINKETRGKIPKLFDEINPETKLILVDYILFKGKWLTPFDPVFTEVDTFHLDKYKTIKVPMMYGAGKFASTFDKNFRCHVLKLPYQGNATMLVVLMEKMGDHLALEDYLTTDLVETWLRNMKTRNMEVFFPKFKLDQKYEMHELLRQMGIRRIFSPFADLSELSATGRNLQVSRVLQRTVIEVDERGTEAVAGILSEITAYSMPPVIKVDRPFHFMIYEETSGMLLFLGRVVNPTLL

Biochem/physiol Actions

Serpin family A member 10 (SERPIN10) inhibits coagulation factors X and XI. Variations in the gene encoding this protein have been associated with thrombophilia.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single Nucleotide Variant rs2232710 in the Protein Z-Dependent Protease Inhibitor (ZPI, SERPINA10) Gene Is Not Associated with Deep Vein Thrombosis.
Gorski MM
PLoS ONE, 11(3), e0151347-e0151347 (2016)
Novel markers for enterochromaffin cells and gastrointestinal neuroendocrine carcinomas
Justyna L, et.al
Modern Pathology, 22, 261-272 (2009)
Down-regulated expression of SERPIN genes located on chromosome 18q21 in oral squamous cell carcinomas
Masashi S, et.al
Oncology Reports, 24(1), 241-249 (2010)
Down-regulated expression of SERPIN genes located on chromosome 18q21 in oral squamous cell carcinomas.
Shiiba M
Oncology Reports, 24(1), 241-249 (2010)
Justyna Leja et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 22(2), 261-272 (2008-10-28)
The gene expression profile of metastasizing serotonin-producing neuroendocrine carcinomas, which arise from enterochromaffin cells in the jejunum and ileum, is still largely unknown. The aim of this study was to identify genes and proteins, which are preferentially expressed by neuroendocrine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service