Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0027019M1

Sigma-Aldrich

Monoclonal Anti-DNAI1 antibody produced in mouse

clone 2D10-2A5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CILD1, Anti-ICS, Anti-MGC26204, Anti-PCD, Anti-dynein, axonemal, intermediate polypeptide 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D10-2A5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DNAI1(27019)

General description

The inner- and outer-arm dyneins, which bridge between the doublet microtubules in axonemes, are the force-generating proteins responsible for the sliding movement in axonemes. The intermediate and light chains, thought to form the base of the dynein arm, help mediate attachment and may also participate in regulating dynein activity. This gene encodes an intermediate chain dynein, belonging to the large family of motor proteins. Mutations in this gene result in abnormal ciliary ultrastructure and function associated with primary ciliary dyskinesia (PCD) and Kartagener syndrome. (provided by RefSeq)

Immunogen

DNAI1 (AAH30583, 1 a.a. ~ 699 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MIPASAKSPHKQPHKQSISIGRGTRKRDEDSGTEVGEGTDEWAQSKATVRPPDQLELTDAELKEEFTRILTANNPHAPQNIVRYSFKEGTYKPIGFVNQLAVHYTQVGNLIPKDSDEGRRQHYRDELVAGSQESVKVISETGNLEEDEEPKELETEPGSQTDVPAAGAAEKVTEEELMTPKQPKERKLTNQFNFSERASQTCNNPVRDRECQTEPPPRTNFSATANQWEIYDAYVEELEKQEKTKEKEKAKTPVAKKSGKMAMRKLTSMESQTDDLIKLSQAAKIMERMVNQNTYDDIAQDFKYYDDAADEYRDQVGTLLPLWKFQNDKAKRLSVTALCWNPKYRDLFAVGYGSYDFMKQSRGMLLLYSLKNPSFPEYMFSSNSGVMCLDIHVDHPYLVAVGHYDGNVAIYNLKKPHSQPSFCSSAKSGKHSDPVWQVKWQKDDMDQNLNFFSVSSDGRIVSWTLVKRKLVHIDVIKLKVEGSTTEVPEGLQLHQVGCGTAFDFHKEIDYMFLVGTEEGKIYKCSKSYSSQFLDTYDAHNMSVDTVSWNPYHTKVFMSCSSDWTVKIWDHTIKTPMFIYDLNSAVGDVAWAPYSSTVFAAVTTDGKAHIFDLAINKYEAICNQPVAAKKNRLTHVQFNLIHPIIIVGDDRGHIISLKLSPNLRKMPKEKKGQEVQKGPAVEIAKLDKLLNLVREVKIKT

Biochem/physiol Actions

DNAI1 (Dynein axonemal intermediate chain 1) encodes an intermediate of outer dynein arms (ODA) proteins of cilia. Mutation in DNAI1 gene causes a trilogy syndrome named as Kartagener syndrome (KS), a rare recessive hereditary disorder named primary ciliary dyskinesia (PCD).

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mike Failly et al.
Respiration; international review of thoracic diseases, 76(2), 198-204 (2008-04-25)
Primary ciliary dyskinesia (PCD) is a rare recessive hereditary disorder characterized by dysmotility to immotility of ciliated and flagellated structures. Its main symptoms are respiratory, caused by defective ciliary beating in the epithelium of the upper airways (nose, bronchi and
Maimoona A Zariwala et al.
American journal of respiratory and critical care medicine, 174(8), 858-866 (2006-07-22)
Primary ciliary dyskinesia (PCD) is a rare, usually autosomal recessive, genetic disorder characterized by ciliary dysfunction, sino-pulmonary disease, and situs inversus. Disease-causing mutations have been reported in DNAI1 and DNAH5 encoding outer dynein arm (ODA) proteins of cilia. We analyzed
C Guichard et al.
American journal of human genetics, 68(4), 1030-1035 (2001-03-07)
Kartagener syndrome (KS) is a trilogy of symptoms (nasal polyps, bronchiectasis, and situs inversus totalis) that is associated with ultrastructural anomalies of cilia of epithelial cells covering the upper and lower respiratory tracts and spermatozoa flagellae. The axonemal dynein intermediate-chain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service