Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0023443M1

Sigma-Aldrich

Monoclonal Anti-SLC35A3 antibody produced in mouse

clone 4B6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DKFZp781P1297, Anti-solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC35A3(23443)

General description

Solute carrier family 35 member A3 (SLC35A3) encodes a uridine diphosphate N-acetylglucosamine transporter, which is localized in the Golgi apparatus.The gene is located on human chromosome 1p21.2.

Immunogen

SLC35A3 (NP_036375, 61 a.a. ~ 113 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DSKCSLRALNRVLHDEILNKPMETLKLAIPSGIYTLQNNLLYVALSNLDAATY

Biochem/physiol Actions

Solute carrier family 35 member A3 (SLC35A3) plays an important role in the glycosylation of macromolecules. The protein plays an important role in skeletal system development. Mutations of this gene is associated with complex vertebral malformation (CVM) in cattle and syndromic autism and epilepsy and arthrogryposis in humans.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic polymorphism of SLC35A3 gene in Karan Fries bull calves
Yathish,, et al.
Indian Journal of Animal Sciences (India) (2011)
Combinations of newly confirmed Glioma-Associated loci link regions on chromosomes 1 and 9 to increased disease risk
Yang, Tu, et al.
BMC Medical Genomics, 4(1), 63-63 (2011)
A human case of SLC35A3-related skeletal dysplasia
Edmondso, et al.
American Journal of Medical Genetics. Part A, 173(10), 2758-2762 (2017)
Recessive mutations in SLC35A3 cause early onset epileptic encephalopathy with skeletal defects
Marini,, et al.
American Journal of Medical Genetics. Part A, 173(4), 1119-1123 (2017)
UDP-N-acetylglucosamine transporter (SLC35A3) regulates biosynthesis of highly branched N-glycans and keratan sulfate
Maszczak, et al.
The Journal of biological chemistry, jbc-M113 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service