Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

WH0022821M1

Sigma-Aldrich

Monoclonal Anti-RASA3 antibody produced in mouse

clone 1F11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-GAP1IP4BP, Anti-GAPIII, Anti-MGC46517, Anti-MGC47588, Anti-RAS p21 protein activator 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... RASA3(22821)

General description

The protein encoded by this gene is member of the GAP1 family of GTPase-activating proteins. The gene product stimulates the GTPase activity of normal RAS p21 but not its oncogenic counterpart. Acting as a suppressor of RAS function, the protein enhances the weak intrinsic GTPase activity of RAS proteins resulting in the inactive GDP-bound form of RAS, thereby allowing control of cellular proliferation and differentiation. This family member is an inositol 1,3,4,5-tetrakisphosphate-binding protein, like the closely related RAS p21 protein activator 2. The two family members have distinct pleckstrin-homology domains, with this particular member having a domain consistent with its localization to the plasma membrane. (provided by RefSeq)

Immunogen

RASA3 (AAH47242, 725 a.a. ~ 834 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DGDRETERIYSLFNLYMSKLEKMQEACGSKSVYDGPEQEEYSTFVIDDPQETYKTLKQVIAGVGALEQEHAQYKRDKFKKTKYGSQEHPIGDKSFQNYIRQQSETSTHSI

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service