Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

WH0011138M2

Sigma-Aldrich

Monoclonal Anti-TBC1D8 antibody produced in mouse

clone 1A12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-AD3, Anti-HBLP1, Anti-TBC1 domain family, member 8 (with GRAM domain), Anti-VRP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A12, monoclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TBC1D8(11138)

General description

TBC1 domain family member 8 (TBC1D8) gene is located on human chromosome 2q11.2.

Immunogen

TBC1D8 (NP_008994, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MEQLADVTLRRLLDNEVFDLDPDLQEPSQITKRDLEARAQNEFFRAFFRLPRKEKLHAVVDCSLWTPFSRCHTAGRMFASDSYICFASREDGCCKIILPLREVVSIEKME

Biochem/physiol Actions

TBC1 domain family member 8 (TBC1D8) is involved in blood coagulation and circulation. It functions as a GTPase-activating protein for the Rab family. The protein is associated with membrane trafficking.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adaptation and possible ancient interspecies introgression in pigs identified by whole-genome sequencing
Ai H, et al.
Nature Genetics, 47(3), 217-217 (2015)
Multiplex image-based autophagy RNAi screening identifies SMCR8 as ULK1 kinase activity and gene expression regulator
Jung J, et al.
eLife, 6(3), e23063-e23063 (2017)
An integration of genome-wide association study and gene expression profiling to prioritize the discovery of novel susceptibility Loci for osteoporosis-related traits
Hsu YH, et al.
PLoS Genetics, 6(6), e1000977-e1000977 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service