Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0010855M5

Sigma-Aldrich

Monoclonal Anti-HPSE antibody produced in mouse

clone 4D7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-HPA, Anti-HPR1, Anti-HPSE1, Anti-HSE1, Anti-heparanase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4D7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HPSE(10855)

General description

The heparanase gene (HPSE) is a member of the Ets gene family. It is expressed in cytotrophoblasts, endothelial cells, keratinocytes, mast cells and hematopoietic cells like platelets, neutrophils, macrophages, T and B lymphocytes. HPSE gene is located on human chromosome 4q21.

Immunogen

HPSE (NP_006656.1, 72 a.a. ~ 170 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LILLGSPKLRTLARGLSPAYLRFGGTKTDFLIFDPKKESTFEERSYWQSQVNQDICKYGSIPPDVEEKLRLEWPYQEQLLLREHYQKKFKNSTYSRSSV

Biochem/physiol Actions

The heparanase gene (HPSE/HPA) helps in the degeneration of heparan chains (HS). It controls the structure and activities of heparan sulfate proteoglycan (HSPG). This gene participates in the differentiation and development of prostate epithelial cells. HPA play a major role in cervical cancer metastasis and angiogenesis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prostate-specific transcription factor hPSE is translated only in normal prostate epithelial cells
Nozawa M, et al.
Cancer Research, 60(5), 1348-1352 (2000)
Association of heparanase gene (HPSE) single nucleotide polymorphisms with hematological malignancies
Ostrovsky O, et al.
Leukemia, 21(11), 2296-2303 (2007)
Novel 1, 3-N, O-Spiroheterocyclic compounds inhibit heparanase activity and enhance nedaplatin-induced cytotoxicity in cervical cancer cells
Song Y, et al.
Oncotarget, 7(24), 36154-36167 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service