Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

WH0010097M1

Sigma-Aldrich

Monoclonal Anti-ACTR2 antibody produced in mouse

clone 1B10-C4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ARP2, Anti-ARP2 actin-related protein 2 homolog (yeast)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$406.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B10-C4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTR2(10097)

General description

ARP2 actin related protein 2 homolog (ACTR2) is encoded by the gene mapped to human chromosome 2p14.
The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion. Two transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

ACTR2 (AAH14546.1, 1 a.a. ~ 394 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDSQGRKVVVCDNGTGFVKCGYAGSNFPEHIFPALVGRPIIRSTTKVGNIEIKDLMVGDEASELRSTLEVNYPMENGIVRNWDDMKHLWDYTFGPEKLNIDTRNCKILLTEPPMNPTKNREKIVEVMFETYQFSGVYVAIQAVLTLYAQGLLTGVVVDSGDGVTHICPVYESFSLPHLTRRLDIAGRDITRYLIKLLLLRGYAFNHSADFETVRMIKEKLCYVGYNIEQEQKLALETTVLVESYTLPDGRIIKVGGERFEAPEALFQPHLINVEGVGVAELLFNTIQAADIDTRSEFYKHIVLSGGSTMYPGLPSRLERELKQLYLERVLKGDVEKLSKFKIRIEDPPRRKHMVFLGGAVLADIMKDKDNFWMTRQEYQEKGVRVLEKLGVTVR

Biochem/physiol Actions

ARP2 actin related protein 2 homolog (ACTR2) is a key component of the Arp2/3 complex. It is required for the regulation of the actin cytoskeleton. It plays a vital role in various processes such as cell locomotion and intracellular motility of lipid vesicles. In addition, It is also involved in the regulation of axon guidance and growth cone filopodia formation in Caenorhabditis elegans and Drosophila.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical and molecular characterization of two patients with overlapping de novo microdeletions in 2p14-p15 and mild mental retardation.
Wohlleber E, et al.
European Journal of Medical Genetics, 54(1), 67-72 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service