Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0009750M1

Sigma-Aldrich

Monoclonal Anti-C6orf32 antibody produced in mouse

clone 2E6-1B10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DIFF40, Anti-DIFF48, Anti-FAM65B, Anti-KIAA0386, Anti-PL48, Anti-chromosome 6 open reading frame 32

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E6-1B10, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAM65B(9750)

Related Categories

General description

FAM65B (family with sequence similarity 65, member B), also known as chromosome 6 open reading frame 32 (C6ORF32), is mapped to human chromosome 6p22.3. FAM65B is known to be expressed during embryonic and postnatal development stages in cochlea. The encoded protein is found to concentrate in the plasma membranes of the stereocilia of inner and outer hair cells of the inner ear.

Immunogen

C6orf32 (AAH01232.1, 1 a.a. ~ 591 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MLVGSQSFSPGGPNGIIRSQSFAGFSGLQERRSRCNSFIENSSALKKPQAKLKKMHNLGHKNNNPPKEPQPKRVEEVYRALKNGLDEYLEVHQTELDKLTAQLKDMKRNSRLGVLYDLDKQIKTIERYMRRLEFHISKVDELYEAYCIQRRLQDGASKMKQAFATSPASKAARESLTEINRSFKEYTENMCTIEVELENLLGEFSIKMKGLAGFARLCPGDQYEIFMKYGRQRWKLKGKIEVNGKQSWDGEETVFLPLIVGFISIKVTELKGLATHILVGSVTCETKELFAARPQVVAVDINDLGTIKLNLEITWYPFDVEDMTASSGAGNKAAALQRRMSMYSQGTPETPTFKDHSFFSNLPDDIFENGKAAEEKMPLSLSFSDLPNGDCALTSHSTGSPSNSTNPEITITPAEFNLSSLASQNEGMDDTSSASSRNSLGEGQEPKSHLKEEDPEEPRKPASAPSEACRRQSSGAGAEHLFLENDVAEALLQESEEASELKPVELDTSEGNITKQLVKRLTSAEVPMATDRLLSEGSVGGESEGCRSFLDGSLEDAFNGLLLALEPHKEQYKEFQDLNQEVMNLDDILKK

Biochem/physiol Actions

FAM65B (family with sequence similarity 65, member B) is recognized as a Ras homolog gene family, member A (RHOA) inhibitor and is involved in polarization. FAM65B regulates RhoA activity with the help of forkhead box O transcription factors. It binds to RhoA and blocks the GTP loading to RhoA. FAM65B deficiency in neutrophils leads to an increase in RhoA activity and causes defects in chemotaxis directionality and adhesion to endothelial cells under flow. It is involved in chemokine-stimulated migration and adhesion of Jurkat cells. Phosphorylation stabilizes FAM65B. FAM65B is a component of the inner ear and is involved in the process of hearing. Mutation in the gene is known to be associated with hearing loss in humans. FAM65B is associated with myotube formation and in the regulation of cell adhesion, polarization, and migration. Overexpression of the protein in myogenic and non-myogenic cells results in the formation of cellular protrusions. Hence, FAM65B might be involved in myoblast migration or cytoskeletal protein rearrangements that accompany myogenic differentiation.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Front-signal-dependent accumulation of the RHOA inhibitor FAM65B at leading edges polarizes neutrophils
Kun Gao
Journal of Cell Science (2015)
Fam65b is important for formation of the HDAC6-dysferlin protein complex during myogenic cell differentiation
Anuradha B
Faseb Journal (2014)
FAM65B is a membrane-associated protein of hair cell stereocilia required for hearing
Oscar Diaz-Horta
Proceedings of the National Academy of Sciences of the USA (2014)
Fam65b is a new transcriptional target of FOXO1 that regulates RhoA signaling for T lymphocyte migration.
Rougerie P
Journal of Immunology (2013)
Soonsang Yoon et al.
Developmental biology, 301(1), 70-81 (2006-12-08)
We have identified a gene by microarray analysis that is located on chromosome 6 (c6orf32), whose expression is increased during human fetal myoblast differentiation. The protein encoded by c6orf32 is expressed both in myogenic and non-myogenic primary cells isolated from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service