Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

WH0009748M1

Sigma-Aldrich

Monoclonal Anti-SLK antibody produced in mouse

clone 3E9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-KIAA0204, Anti-STE20-like kinase (yeast), Anti-STK2, Anti-bA16H23.1, Anti-se209

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E9, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLK(9748)

General description

STE20 like kinase (SLK) is a 1204 amino acid serine/threonine kinase. It possesses a kinase domain located at the amino-terminal and a C-terminal regulatory domain with coiled-coils. SLK is group V germinal center kinase that is expressed in the nucleus, cytosol, microtubules and centrosomes. The SLK gene is localized on human chromosome 10q24.33.

Immunogen

SLK (NP_055535, 1126 a.a. ~ 1235 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KHENQMRDLQLQCEANVRELHQLQNEKCHLLVEHETQKLKELDEEHSQELKEWREKLRPRKKTLEEEFARKLQEQEVFFKMTGESECLNPSTQSRISKFYPIPSLHSTGS

Biochem/physiol Actions

STE20 like kinase (SLK) is a modulator of cell motility. It phosphorylates cytoskeleton-bound proteins, which take part in the remodeling of migratory cell architecture. SLK is also involved in mitogen-activated protein kinase pathways. It acts as an activator of Jun N-terminal kinase (JNK) and transactivates tumor suppressor p53.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SLK/LOSK kinase regulates cell motility independently of microtubule organization and Golgi polarization.
Fokin A I, et al.
Cytoskeleton (Hoboken, N.J.), 73(2), 83-92 (2016)
Molecular cloning and characterization of a novel human STE20-like kinase, hSLK.
Yamada E, et al.
Biochimica et Biophysica Acta, 1495(3), 250-262 (2000)
BET1L and TNRC6B associate with uterine fibroid risk among European Americans.
Todd L E, et al.
Human Genetics, 132(8), 943?953-943?953 (2013)
Regulation of the Ste20-like kinase, SLK: involvement of activation segment phosphorylation.
Luhovy A Y, et al.
The Journal of Biological Chemistry, 287(8), 5446-5458 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service