Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

WH0009457M1

Sigma-Aldrich

Monoclonal Anti-FHL5 antibody produced in mouse

clone 1G12-D2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ACT, Anti-FLJ33049, Anti-KIAA0776, Anti-dJ393D12.2, Anti-four and a half LIM domains 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$406.00

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$406.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G12-D2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FHL5(9457)

General description

The protein encoded by this gene is coordinately expressed with activator of cAMP-responsive element modulator (CREM). It is associated with CREM and confers a powerful transcriptional activation function. CREM acts as a transcription factor essential for the differentiation of spermatids into mature spermatozoa. There are multiple polyadenylation sites found in this gene. (provided by RefSeq)

Immunogen

FHL5 (AAH21723, 1 a.a. ~ 284 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MTTAHFYCQYCTASLLGKKYVLKDDSPYCVTCYDRVFSNYCEECKKPIESDSKDLCYKDRHWHEGCFKCTKCNHSLVEKPFAAKDERLLCTECYSNECSSKCFHCKRTIMPGSRKMEFKGNYWHETCFVCENCRQPIGTKPLISKESGNYCVPCFEKEFAHYCNFCKKVITSGGITFCDQLWHKECFLCSDCRKDLCEEQFMSRDDYPFCMDCYNHLYANKCVACSKPISGLTGAKFICFQDSQWHSECFNCGKCSVSLVGKGFLTQNKEIFCQKCGSGMDTDI

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service